2-Amino-6-fluoro-3-nitrobenzotrifluoride

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-6-nitro-2-(trifluoromethyl)aniline, 6-Nitro-α,α,α,3-tetrafluoro-o-toluidine

PC49364-1G PC49364-250MG PC49364-5G
APOSPC49364-1GEA 296 GBP
APOSPC49364-1G APOSPC49364-250MG APOSPC49364-5G
2-Amino-6-fluoro-3-nitrobenzotrifluoride
2-Amino-6-fluoro-3-nitrobenzotrifluoride
Formula: C₇H₄F₄N₂O₂
MW: 224,112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24850038
CAS Number: 1440535-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR