2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Iodo-6-nitro-4-[(trifluoromethyl)sulphanyl]phenol, 4-Hydroxy-3-iodo-5-nitrophenyl trifluoromethyl sulphide
PC49360-1G
PC49360-250MG
APOSPC49360-1GEA
117
GBP
APOSPC49360-1G
APOSPC49360-250MG
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol
Formula:
C₇H₃F₃INO₃S MW: 365,068 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24850032 CAS Number: 1440535-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...