6-Bromo-4-(trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole 97%

Supplier: Apollo Scientific
Warning

PC49316-1G PC49316-250MG
APOSPC49316-1GEA 420 GBP
APOSPC49316-1G APOSPC49316-250MG
6-Bromo-4-(trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole 97%
6-Bromo-4-(trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole
Formula: C₉H₃BrF₆N₂O
MW: 349,027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628767
CAS Number: 1417341-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR