2-Bromo-6-(difluoromethyl)pyridine 95+%

Supplier: Apollo Scientific
Danger

Synonyms: 6-Bromo-α,α-difluoro-2-picoline

PC49145-10G PC49145-1G PC49145-5G
APOSPC49145-10GEA 257 GBP
APOSPC49145-10G APOSPC49145-1G APOSPC49145-5G
2-Bromo-6-(difluoromethyl)pyridine 95+%
2-Bromo-6-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
MW: 208,003 g/mol
Density: 1,7025 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD16140194
CAS Number: 872365-91-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR