2-(4-Amino-3-methyl-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
Supplier: Apollo Scientific
|
Warning
|
PC48792-1G
PC48792-250MG
APOSPC48792-1GEA
551
GBP
APOSPC48792-1G
APOSPC48792-250MG
2-(4-Amino-3-methyl-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(4-Amino-3-methyl-5-nitrophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formula:
C₁₀H₈F₆N₂O₃ MW: 318,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31722922 CAS Number: 2366994-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...