5-Bromo-2-chloro-4-fluoronitrobenzene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-4-chloro-2-fluoro-5-nitrobenzene

PC48769-1G PC48769-5G PC48769-10G
APOSPC48769-1GEA 44.6 GBP
APOSPC48769-1G APOSPC48769-5G APOSPC48769-10G
5-Bromo-2-chloro-4-fluoronitrobenzene 97%
5-Bromo-2-chloro-4-fluoronitrobenzene
Formula: C₆H₂BrClFNO₂
MW: 254,441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024826
CAS Number: 1311197-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR