2,2-Difluoroprop-1-yl toluene-4-sulphonate

Supplier: Apollo Scientific
Warning

Synonyms: 2,2-Difluoroprop-1-yl tosylate, 2,2-Difluoroprop-1-yl 4-methylbenzenesulphonate

PC48442-1G PC48442-250MG
APOSPC48442-1GEA 231 GBP
APOSPC48442-1G APOSPC48442-250MG
2,2-Difluoroprop-1-yl toluene-4-sulphonate
2,2-Difluoroprop-1-yl toluene-4-sulphonate
Formula: C₁₀H₁₂F₂O₃S
MW: 250,262 g/mol
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD20489424
CAS Number: 1262400-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR