N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide
Supplier: Apollo Scientific
|
Warning
|
PC450563-1G
PC450563-250MG
APOSPC450563-1GEA
746
GBP
APOSPC450563-1G
APOSPC450563-250MG
N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide
N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide
Formula:
C₁₂H₁₁F₅N₂O₂ MW: 310,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30062918 CAS Number: 1980048-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...