Bis(2,2-difluoroethyl) carbonate
Supplier: Apollo Scientific
|
Warning
|
PC450292-1G
PC450292-5G
APOSPC450292-1GEA
125
GBP
APOSPC450292-1G
APOSPC450292-5G
Bis(2,2-difluoroethyl) carbonate
Bis(2,2-difluoroethyl) carbonate
Formula:
C₅H₆F₄O₃ MW: 190,093 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28334287 CAS Number: 916678-16-5 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...