1-(Nonafluorobutyl)trifluoroethanesultone 97%

Supplier: Apollo Scientific
Warning

PC450248-5G PC450248-1G
APOSPC450248-5GEA 171 GBP
APOSPC450248-5G APOSPC450248-1G
1-(Nonafluorobutyl)trifluoroethanesultone 97%
1-(Nonafluorobutyl)trifluoroethanesultone
Formula: C₆F₁₂O₃S
MW: 380.108 g/mol
Boiling Pt: 114 °C (76 mm Hg)
Density: 1.78 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD16621321
CAS Number: 85211-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR