3-(2-Chloroethoxy)-1,1,2,2-tetrafluoropropane

Supplier: Apollo Scientific
Warning

PC450226-1G PC450226-5G
APOSPC450226-1GEA 98.3 GBP
APOSPC450226-1G APOSPC450226-5G
3-(2-Chloroethoxy)-1,1,2,2-tetrafluoropropane
3-(2-Chloroethoxy)-1,1,2,2-tetrafluoropropane
Formula: C₅H₇ClF₄O
MW: 194,555 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11164793
CAS Number: 64769-39-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR