2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane
Supplier: Apollo Scientific
PC450206-1G
PC450206-5G
APOSPC450206-1GEA
125
GBP
APOSPC450206-1G
APOSPC450206-5G
2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane
2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane
Formula:
C₅H₂F₉I MW: 359,959 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074329 CAS Number: 428-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...