Sodium 2,4,5-trifluorophenylacetate

Supplier: Apollo Scientific

PC450165-1G PC450165-5G
APOSPC450165-1GEA 125 GBP
APOSPC450165-1G APOSPC450165-5G
Sodium 2,4,5-trifluorophenylacetate
Sodium 2,4,5-trifluorophenylacetate
Formula: C₈H₄F₃O₂·Na
MW: 212.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054314
CAS Number: 1980063-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR