Tris(3-fluoropropyl)phosphate

Supplier: Apollo Scientific
Warning

PC450091-5G PC450091-1G
APOSPC450091-5GEA 414 GBP
APOSPC450091-5G APOSPC450091-1G
Tris(3-fluoropropyl)phosphate
Tris(3-fluoropropyl)phosphate
Formula: C₉H₁₈F₃O₄P
MW: 278,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20489415
CAS Number: 1346521-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR