2-Chloro-4-(trifluoromethyl)benzamide 95+%
Supplier: Apollo Scientific
PC446190-1G
PC446190-5G
APOSPC446190-1GEA
129
GBP
APOSPC446190-1G
APOSPC446190-5G
2-Chloro-4-(trifluoromethyl)benzamide 95+%
2-Chloro-4-(trifluoromethyl)benzamide
Formula:
C₈H₅ClF₃NO MW: 223,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01571094 CAS Number: 254880-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...