3-(Chloromethyl)-2-methyl-6-(trifluoromethyl)pyridine 95+%
Supplier: Apollo Scientific
PC446105-1G
PC446105-500MG
PC446105-5G
APOSPC446105-1GEA
239
GBP
APOSPC446105-1G
APOSPC446105-500MG
APOSPC446105-5G
3-(Chloromethyl)-2-methyl-6-(trifluoromethyl)pyridine 95+%
3-(Chloromethyl)-2-methyl-6-(trifluoromethyl)pyridine
Formula:
C₈H₇ClF₃N MW: 209,596 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189686 CAS Number: 917396-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...