6-Methyl-4-trifluoromethylpyridine-2-carboxaldehyde 95+%

Supplier: Apollo Scientific

PC446104-1G PC446104-500MG
APOSPC446104-1GEA 477 GBP
APOSPC446104-1G APOSPC446104-500MG
6-Methyl-4-trifluoromethylpyridine-2-carboxaldehyde 95+%
6-Methyl-4-trifluoromethylpyridine-2-carboxaldehyde
Formula: C₈H₆F₃NO
MW: 189,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261838
CAS Number: 451459-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR