3,4-Difluorobenzimidic acid ethyl ester hydrochloride 95+%
Supplier: Apollo Scientific
PC446097-1G
PC446097-500MG
PC446097-5G
APOSPC446097-1GEA
239
GBP
APOSPC446097-1G
APOSPC446097-500MG
APOSPC446097-5G
3,4-Difluorobenzimidic acid ethyl ester hydrochloride 95+%
3,4-Difluorobenzimidic acid ethyl ester hydrochloride
Formula:
C₉H₉F₂NO·ClH MW: 221,632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12131139 CAS Number: 1205638-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...