2-Chloro-N-(6-chloro-4-trifluoromethylpyridin-2-yl)acetamide 95+%

Supplier: Apollo Scientific

PC446093-1G PC446093-500MG
APOSPC446093-1GEA 372 GBP
APOSPC446093-1G APOSPC446093-500MG
2-Chloro-N-(6-chloro-4-trifluoromethylpyridin-2-yl)acetamide 95+%
2-Chloro-N-(6-chloro-4-trifluoromethylpyridin-2-yl)acetamide
Formula: C₈H₅Cl₂F₃N₂O
MW: 273,039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981216
CAS Number: 1311279-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR