2-Methyl-5-(trifluoromethyl)-2H-pyrazole-3-thiol 95+%

Supplier: Apollo Scientific

PC446091-1G PC446091-500MG
APOSPC446091-1GEA 286 GBP
APOSPC446091-1G APOSPC446091-500MG
2-Methyl-5-(trifluoromethyl)-2H-pyrazole-3-thiol 95+%
2-Methyl-5-(trifluoromethyl)-2H-pyrazole-3-thiol
Formula: C₅H₅F₃N₂S
MW: 182,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845724
CAS Number: 191334-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR