(4-Fluorophenyl)morpholin-4-ylacetonitrile hydrochloride 95+%

Supplier: Apollo Scientific

PC446088-10G PC446088-1G PC446088-5G
APOSPC446088-10GEA 533 GBP
APOSPC446088-10G APOSPC446088-1G APOSPC446088-5G
(4-Fluorophenyl)morpholin-4-ylacetonitrile hydrochloride 95+%
(4-Fluorophenyl)morpholin-4-ylacetonitrile hydrochloride
Formula: C₁₂H₁₃FN₂O·ClH
MW: 256,704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052437
CAS Number: 1208081-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR