6-Fluoro-1H-indazole-3-carbonitrile 95%
Supplier: Apollo Scientific
PC421225-1G
PC421225-5G
APOSPC421225-1GEA
211
GBP
APOSPC421225-1G
APOSPC421225-5G
6-Fluoro-1H-indazole-3-carbonitrile 95%
6-Fluoro-1H-indazole-3-carbonitrile
Formula:
C₈H₄FN₃ MW: 161,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371570 CAS Number: 885278-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...