3-Amino-5-fluorophenylboronic acid

Supplier: Apollo Scientific

PC412218-1G PC412218-250MG
APOSPC412218-1GEA 584 GBP
APOSPC412218-1G APOSPC412218-250MG
3-Amino-5-fluorophenylboronic acid
3-Amino-5-fluorophenylboronic acid
Formula: C₆H₇BFNO₂
MW: 154,935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025966
CAS Number: 1150114-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR