2,2-Difluorocyclopenta-1,3-dione

Supplier: Apollo Scientific
Warning

PC409034-1G PC409034-250MG PC409034-500MG
APOSPC409034-1GEA 867 GBP
APOSPC409034-1G APOSPC409034-250MG APOSPC409034-500MG
2,2-Difluorocyclopenta-1,3-dione
2,2-Difluorocyclopenta-1,3-dione
Formula: C₅H₄F₂O₂
Melting Pt: 72…73 °C
Storage Temperature: Ambient
MDL Number: MFCD16657105
CAS Number: 1215071-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR