1-Bromo-3,4,4,4-tetrafluoro-3-(trifluoromethyl)butane

Supplier: Apollo Scientific
Warning

PC408201-25G PC408201-100G PC408201-5G
APOSPC408201-25GEA 159 GBP
APOSPC408201-25G APOSPC408201-100G APOSPC408201-5G
1-Bromo-3,4,4,4-tetrafluoro-3-(trifluoromethyl)butane
1-Bromo-3,4,4,4-tetrafluoro-3-(trifluoromethyl)butane
Formula: C₅H₄BrF₇
Storage Temperature: Refrigerator
MDL Number: MFCD09265345
CAS Number: 1274903-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR