Iododifluoroacetic acid 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,2-Difluoro-2-iodoacetic acid
PC408091-5G
PC408091-250MG
PC408091-1G
APOSPC408091-5GEA
342
GBP
APOSPC408091-5G
APOSPC408091-250MG
APOSPC408091-1G
Iododifluoroacetic acid 95%
Iododifluoroacetic acid
Formula:
C₂HF₂IO₂ MW: 221,929 g/mol Boiling Pt: 82…87 °C (45 mm Hg) Melting Pt: 92…94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD27918484 CAS Number: 1032021-83-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...