3-Fluorobicyclo[1.1.1]pentane-1-carboxamide
Supplier: Apollo Scientific
PC405699-1G
PC405699-250MG
APOSPC405699-1GEA
2170
GBP
APOSPC405699-1G
APOSPC405699-250MG
3-Fluorobicyclo[1.1.1]pentane-1-carboxamide
3-Fluorobicyclo[1.1.1]pentane-1-carboxamide
Formula:
C₆H₈FNO MW: 129,132 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19207132 CAS Number: 146038-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...