2-Fluorobenzoyl isocyanate

Supplier: Apollo Scientific

PC404019-1G PC404019-250MG
APOSPC404019-1GEA 460 GBP
APOSPC404019-1G APOSPC404019-250MG
2-Fluorobenzoyl isocyanate
2-Fluorobenzoyl isocyanate

May form small amount of sediment

Formula: C₈H₄FNO₂
MW: 165,121 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16301148
CAS Number: 62869-34-5
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR