2-Bromo-5-fluoropyridin-3-ol 95%
Supplier: Apollo Scientific
|
Warning
|
PC400188-1G
PC400188-250MG
APOSPC400188-1GEA
91.6
GBP
APOSPC400188-1G
APOSPC400188-250MG
2-Bromo-5-fluoropyridin-3-ol 95%
2-Bromo-5-fluoropyridin-3-ol
Formula:
C₅H₃BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD11519042 CAS Number: 1093758-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...