4-Ethynyl-3-fluoroaniline

Supplier: Apollo Scientific
Warning

PC400177-1G PC400177-250MG
APOSPC400177-1GEA 216 GBP
APOSPC400177-1G APOSPC400177-250MG
4-Ethynyl-3-fluoroaniline
4-Ethynyl-3-fluoroaniline
Formula: C₈H₆FN
MW: 135,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036455
CAS Number: 1233501-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR