1-(2-Bromoethynyl)-2-fluorobenzene
Supplier: Apollo Scientific
|
Danger
|
PC400026-250MG
PC400026-500MG
APOSPC400026-250MGEA
314
GBP
APOSPC400026-250MG
APOSPC400026-500MG
1-(2-Bromoethynyl)-2-fluorobenzene
1-(2-Bromoethynyl)-2-fluorobenzene
Formula:
C₈H₄BrF MW: 199.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31536744 CAS Number: 1246224-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...