3-[(Trifluoromethyl)sulphonyloxy]pyridazine

Supplier: Apollo Scientific

PC3316-100MG PC3316-250MG
APOSPC3316-100MGEA 216 GBP
APOSPC3316-100MG APOSPC3316-250MG
3-[(Trifluoromethyl)sulphonyloxy]pyridazine
3-[(Trifluoromethyl)sulphonyloxy]pyridazine
Formula: C₅H₃F₃N₂O₃S
MW: 228,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461637
CAS Number: 1031928-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR