Ethyl 7-bromo-5-fluorobenzofuran-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
PC32791-1G
PC32791-250MG
APOSPC32791-1GEA
520
GBP
APOSPC32791-1G
APOSPC32791-250MG
Ethyl 7-bromo-5-fluorobenzofuran-2-carboxylate
Ethyl 7-bromo-5-fluorobenzofuran-2-carboxylate
Formula:
C₁₁H₈BrFO₃ MW: 287,082 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31812096 CAS Number: 1259929-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...