5-Bromo-2-methyl-3-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
PC303587-1G
PC303587-5G
APOSPC303587-1GEA
374
GBP
APOSPC303587-1G
APOSPC303587-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzyl alcohol 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzyl alcohol
Formula:
C₉H₈BrF₃O MW: 269.058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27942785 CAS Number: 1266728-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...