5-Bromo-2-methyl-3-(trifluoromethyl)benzaldehyde 97%

Supplier: Apollo Scientific

PC303586-1G PC303586-5G
APOSPC303586-1GEA 253 GBP
APOSPC303586-1G APOSPC303586-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzaldehyde 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzaldehyde
Formula: C₉H₆BrF₃O
MW: 267,043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640162
CAS Number: 2384835-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR