3-Bromo-2,5,6-trifluorophenylacetic acid 97%

Supplier: Apollo Scientific

PC303510-1G PC303510-5G
APOSPC303510-1GEA 377 GBP
APOSPC303510-1G APOSPC303510-5G
3-Bromo-2,5,6-trifluorophenylacetic acid 97%
3-Bromo-2,5,6-trifluorophenylacetic acid
Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28746951
CAS Number: 1805143-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR