3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
PC303465-1G
PC303465-5G
APOSPC303465-1GEA
210
GBP
APOSPC303465-1G
APOSPC303465-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₅BrF₄O MW: 273,022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30834069 CAS Number: 2092564-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...