2-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%

Supplier: Apollo Scientific

PC303381-1G PC303381-250MG
APOSPC303381-1GEA 525 GBP
APOSPC303381-1G APOSPC303381-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzamide
Formula: C₉H₇F₄NO
MW: 221,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31916394
CAS Number: 2384846-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR