2',3'-Difluoro-5'-methoxy-4'-methylacetophenone 97%

Supplier: Apollo Scientific

PC303240-1G PC303240-250MG
APOSPC303240-1GEA 886 GBP
APOSPC303240-1G APOSPC303240-250MG
2',3'-Difluoro-5'-methoxy-4'-methylacetophenone 97%
2',3'-Difluoro-5'-methoxy-4'-methylacetophenone
Formula: C₁₀H₁₀F₂O₂
MW: 200,182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986846
CAS Number: 1706436-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR