2-Fluoro-4-methyl-3-(trifluoromethyl)aniline 97%

Supplier: Apollo Scientific
Warning

PC303214-1G PC303214-250MG
APOSPC303214-1GEA 763 GBP
APOSPC303214-1G APOSPC303214-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)aniline 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)aniline
Formula: C₈H₇F₄N
MW: 193,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20725977
CAS Number: 1000590-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR