2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetonitrile 97%

Supplier: Apollo Scientific

PC303194-1G PC303194-250MG
APOSPC303194-1GEA 662 GBP
APOSPC303194-1G APOSPC303194-250MG
2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetonitrile 97%
2-Fluoro-5-methyl-3-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₇F₄N
MW: 217,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054190
CAS Number: 1706438-79-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR