2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
PC303169-1G
PC303169-250MG
APOSPC303169-1GEA
559
GBP
APOSPC303169-1G
APOSPC303169-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl alcohol 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl alcohol
Formula:
C₉H₈F₄O MW: 208.153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28103363 CAS Number: 1824048-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...