4-Fluoro-3-methyl-2-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
PC303160-5G
PC303160-1G
APOSPC303160-5GEA
488
GBP
APOSPC303160-5G
APOSPC303160-1G
4-Fluoro-3-methyl-2-(trifluoromethyl)aniline 97%
4-Fluoro-3-methyl-2-(trifluoromethyl)aniline
Formula:
C₈H₇F₄N MW: 193,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27986857 CAS Number: 1511180-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...