2-Methoxy-4,6-bis(trifluoromethyl)benzamide 98%
Supplier: Apollo Scientific
PC303038-5G
PC303038-1G
APOSPC303038-5GEA
185
GBP
APOSPC303038-5G
APOSPC303038-1G
2-Methoxy-4,6-bis(trifluoromethyl)benzamide 98%
2-Methoxy-4,6-bis(trifluoromethyl)benzamide
Formula:
C₁₀H₇F₆NO₂ MW: 287,158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00729121 CAS Number: 259655-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...