5-Chloro-2-(trifluoromethoxy)phenylacetonitrile 98%

Supplier: Apollo Scientific

PC302890-1G PC302890-5G
APOSPC302890-1GEA 215 GBP
APOSPC302890-1G APOSPC302890-5G
5-Chloro-2-(trifluoromethoxy)phenylacetonitrile 98%
5-Chloro-2-(trifluoromethoxy)phenylacetonitrile
Formula: C₉H₅ClF₃NO
MW: 235,59 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD11519359
CAS Number: 1092461-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR