4-Ethoxy-3,5-difluorobenzoyl chloride
Supplier: Apollo Scientific
PC302876-10G
PC302876-5G
APOSPC302876-10GEA
296
GBP
APOSPC302876-10G
APOSPC302876-5G
4-Ethoxy-3,5-difluorobenzoyl chloride
4-Ethoxy-3,5-difluorobenzoyl chloride
Formula:
C₉H₇ClF₂O₂ MW: 220,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258684 CAS Number: 1017779-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...