4-Chloro-3-fluoro-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
PC302869-10G
PC302869-5G
PC302869-1G
APOSPC302869-10GEA
418
GBP
APOSPC302869-10G
APOSPC302869-5G
APOSPC302869-1G
4-Chloro-3-fluoro-5-(trifluoromethyl)benzaldehyde 97%
4-Chloro-3-fluoro-5-(trifluoromethyl)benzaldehyde
Formula:
C₈H₃ClF₄O MW: 226,555 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23795332 CAS Number: 1431329-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...