2-Methoxy-5-(pentafluorosulphur)benzamide 97%
Supplier: Apollo Scientific
PC302811-1G
PC302811-5G
PC302811-10G
APOSPC302811-1GEA
315
GBP
APOSPC302811-1G
APOSPC302811-5G
APOSPC302811-10G
2-Methoxy-5-(pentafluorosulphur)benzamide 97%
2-Methoxy-5-(pentafluorosulphur)benzamide
Formula:
C₈H₈F₅NO₂S MW: 277,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27986830 CAS Number: 1706430-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...