3-Fluoro-5-methyl-DL-phenylglycine
Supplier: Apollo Scientific
PC302556-1G
PC302556-5G
PC302556-10G
APOSPC302556-1GEA
144
GBP
APOSPC302556-1G
APOSPC302556-5G
APOSPC302556-10G
3-Fluoro-5-methyl-DL-phenylglycine
3-Fluoro-5-methyl-DL-phenylglycine
Formula:
C₉H₁₀FNO₂ MW: 183,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02662466 CAS Number: 199327-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...