2,4,6-Trifluoro-DL-phenylalanine
Supplier: Apollo Scientific
PC302435-1G
PC302435-5G
APOSPC302435-1GEA
106
GBP
APOSPC302435-1G
APOSPC302435-5G
2,4,6-Trifluoro-DL-phenylalanine
2,4,6-Trifluoro-DL-phenylalanine
Formula:
C₉H₈F₃NO₂ MW: 219,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116038 CAS Number: 1259973-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...